Learn More
Apexbio Technology LLC Dovitinib Dilactic acid 852433-84-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A84185.1
Encompass_Preferred
Dovitinib Dilactic acid (CAS 852433-84-2) is a small molecule inhibitor that targets multiple receptor tyrosine kinases including FLT-3 c-KIT vascular endothelial growth factor receptors (VEGFR) platelet-derived growth factor receptor beta (PDGFR ) and colony-stimulating factor 1 receptor (CSF-1R) By inhibiting these signaling pathways Dovitinib Dilactic acid modulates cellular processes such as proliferation survival and angiogenesis This compound is utilized in biomedical research to investigate the roles of diverse growth factor receptors in oncogenic signaling and tumor microenvironment interactions
Catalog No. 50-000-94484