Learn More
Medchemexpress LLC Dowicil 200 | 51229-78-8 | 98.9% | 251.16 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW009497
Encompass_Preferred
Dowicil 200 (Quaternium-15) is a skin toxin and allergen that inhibits the growth of *Pinctada fucata* cells. It possesses antimicrobial activity, being more potent against fungi and molds than against other fungi.
  • Acts as a skin toxin and allergen.
  • Inhibits the growth of *Pinctada fucata* cells.
  • Exhibits antimicrobial activity.
  • More potent against fungi and molds.
Catalog No. 50-004-51573