Learn More
TARGETMOL CHEMICALS INC Doxapram hydrochloride hydrate
SDP
Supplier:  TARGETMOL CHEMICALS INC T0038200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Doxapram hydrochloride hydrate (Doxapram HCl) effectively inhibits the function of TASK-1, TASK-3, and TASK-1/TASK-3 heterodimeric channels, displaying EC50 values of 410 nM, 37 uM, and 9 uM, respectively. Purity 99.64%
Catalog No. 50-288-0557