Learn More
Selleck Chemical LLC Doxazosin
SDP
Supplier:  Selleck Chemical LLC S57821G
Encompass_Preferred
Doxazosin(UK 33274) is a quinazoline-derivative that selectively antagonizes postsynaptic 1-adrenergic receptors
Catalog No. 50-000-03056