Learn More
Apexbio Technology LLC Doxazosin 74191-85-8 100mg
SDP
Supplier:  Apexbio Technology LLC B1209100
Encompass_Preferred
Doxazosin (CAS 74191-85-8) is a synthetic quinazoline derivative that functions as a selective antagonist of postsynaptic 1-adrenergic receptors By inhibiting these receptors doxazosin reduces vascular smooth muscle contraction leading to decreased peripheral resistance This pharmacological profile is valuable for research investigating 1-adrenergic signaling pathways vascular tone regulation and cardiovascular physiology Doxazosin is commonly utilized in studies related to hypertension benign prostatic hyperplasia and receptor pharmacology
Catalog No. 50-000-90356