Learn More
Medchemexpress LLC Doxazosin | 74191-85-8 | 99.9% | C23H25N5O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB009810MM/1ML
Encompass_Preferred
Doxazosin (UK 33274) is a quinazoline-derivative that selectively antagonizes postsynaptic α1-adrenergic receptors. It is for research use only and not sold to patients.
  • Selectively antagonizes postsynaptic α1-adrenergic receptors
  • Functions as a long-lasting inhibitor of α1-adrenoceptors
  • Used to treat benign prostatic hyperplasia and lower urinary tract symptoms
  • May directly inhibit cholesterol synthesis independently of the LDL receptor
  • Shown effectiveness in monotherapy and combined therapy with a beta-blocker in clinical observations
  • Exhibits minor and transient adverse reactions in some cases
Catalog No. 50-004-32309