Learn More
Selleck Chemical LLC Doxazosin Mesylate S1324-50mg
SDP
Supplier:  Selleck Chemical LLC S132450MG
Encompass_Preferred
Doxazosin a quinazoline-derivative selectively antagonizes postsynaptic 1-adrenergic receptors used in the treatment of high blood pressure and urinary retention associated with benign prostatic hyperplasia
Catalog No. 50-311-1001