Learn More
Medchemexpress LLC Doxazosin mesylate | 77883-43-3 | 98.5% | C24H29N5O8S | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0098A10MM/1ML
Encompass_Preferred
Doxazosin mesylate is a quinazoline-derivative that selectively antagonizes postsynaptic α1-adrenergic receptors. It is the mesylate salt form of doxazosin, a long-lasting inhibitor of α1-adrenoceptors widely used to treat benign prostatic hyperplasia and lower urinary tract symptoms. It may also have a direct inhibitory effect on cholesterol synthesis, potentially leading to upregulation of the LDL receptor and a reduction in LDL cholesterol.
  • Selectively antagonizes postsynaptic α1-adrenergic receptors
  • Long-lasting inhibitor of α1-adrenoceptors
  • Used to treat benign prostatic hyperplasia and lower urinary tract symptoms
  • May have a direct inhibitory effect on cholesterol synthesis
Catalog No. 50-004-33531