Learn More
Abcam Do x azosin mesylate, alpha1 adrenoceptor antagonist, 50MG
Supplier:  Abcam AB12075450MG
Encompass_Preferred
MW 547.58 g/mol, Purity >99%. Selective α₁ adrenoceptor antagonist (pKi values are 9.0, 8.5 and 8.4 at α₁B, α₁A and α₁D receptors, respectively). Shows antihypertensive activity and induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7308