Learn More
Medchemexpress LLC Doxercalciferol (1.alpha.-hydroxyvitamin D2) | 54573-75-0 | 99.9% | 412.65 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY32348100MG
Encompass_Preferred
Doxercalciferol is a Vitamin D2 analog that acts as an activator of the Vitamin D receptor and helps prevent renal disease.
  • Vitamin D2 analog
  • Activator of vitamin D receptor
  • Helps prevent renal disease
  • Elevates serum phosphorus, calcium, and Ca x P in 5/6 nephrectomized rats
  • Blocks PTH from rising and lowers serum PTH
  • Improves proteinuria, prevents podocyte loss, and accumulation of extracellular matrix proteins in HF diet-induced mice
  • Inhibits profibrotic growth factors (TGF-β, PAI-1, CTGF)
  • Suppresses macrophage infiltration, decreases NF-κb activity, and prevents expression of proinflammatory cytokine and renal lipid accumulation
  • Prevents albuminuria, attenuates podocyte loss and apoptosis, and reduces glomerular fibrosis in diabetic mice
Catalog No. 50-004-00659