Learn More
U.S. Pharmacopeia Doxercalciferol, 54573-75-0, MFCD00871065, 2 x 20mg
Supplier:  U.S. Pharmacopeia 1225601
Encompass
Molecular formula C28H44O2, Molecular weight 412.65, Melting point/freezing point 296.6 - 307.4 °F (147 - 153 °C), Synonyms: 1alpha-Hydroxyergocalciferol, 1alpha-Hydroxyvitamin D2, (5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1’,3’-diol
Catalog No. NC1351880