Learn More
Medchemexpress LLC Doxifluridine | 3094-09-5 | 99.4% | C9H11FN2O5 | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0021100G
Encompass_Preferred
Doxifluridine is a 5-FU prodrug that exhibits anticancer activity. It functions as an inhibitor of thymidine synthase and can enhance tumor inhibition through synergism with various drugs.
  • Has anticancer activity
  • Functions as a 5-FU prodrug
  • Inhibits thymidine synthase
  • Enhances tumor inhibition through synergism with other drugs
  • Inhibits angiogenesis by significantly inhibiting VEGF expression in FU-MMT-1 cells
  • Inhibits cell proliferation in HUVEC cells
Catalog No. 50-004-33421