Learn More
Apexbio Technology LLC Doxofylline 69975-86-6 50mg
SDP
Supplier:  Apexbio Technology LLC B173450
Encompass_Preferred
Doxofylline (CAS 69975-86-6) is a synthetic xanthine derivative that acts as a selective phosphodiesterase inhibitor By inhibiting phosphodiesterase enzymes doxofylline increases intracellular cyclic AMP levels leading to bronchodilation Unlike other xanthine compounds it exhibits reduced cardiostimulatory activity and a distinct pharmacological profile Doxofylline is widely studied for its potential to alleviate bronchoconstriction and inflammation in preclinical models of respiratory diseases including asthma making it a valuable tool for investigating airway smooth muscle signaling and respiratory pharmacology
Catalog No. 50-224-0615