Learn More
Sigma Aldrich Fine Chemicals Biosciences Doxorubicin hydrochloride, 25316-40-9, MFCD00077757, 10 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D151510MG
Encompass
Empirical Formula (Hill Notation): C27H29NO11 · HCl, Molecular Weight: 579.98, 98.0-102.0% (HPLC), Synonym: DOX, Hydroxydaunorubicin hydrochloride. Doxorubicin hydrochloride has been used: in cell viability assays, MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide) assay, and as a drug in polymeric PLGA-based microparticulate drug delivery.
Catalog No. NC0916603