Learn More
Cayman Chemical Doxorubicin (hydrochloride), 25316-40-9, 10 mg
SDP
Supplier:  Cayman Chemical 1500710
Encompass_Preferred
Molecular Formula C27H29NO11 • HCl, Purity ≥98%, Formula Weight 580.0, Synonyms: DOX; An anthracycline antitumor antibiotic agent that inhibits DNA topoisomerase II by inducing double-stranded DNA breaks; inhibits nucleic acid synthesis and induces apoptosis by intercalating within DNA.
Catalog No. NC0763727