Learn More
Medchemexpress LLC Doxorubicin-SMCC 100mg | 400647-59-8 | 100 MG

Supplier: Medchemexpress LLC HY116063100MG
Doxorubicin-SMCC is a doxorubicin-based antibody-drug conjugate linker-drug conjugate designed for ADC research, pairing a DNA topoisomerase II inhibitor payload with a non-cleavable SMCC linker for stable conjugation and in vitro studies.
- Contains an SMCC non-cleavable linker for stable thiol coupling.
- Delivers doxorubicin, a DNA topoisomerase II inhibitor, as the cytotoxic payload.
- Purity: 98.8%.
- Formula: C39H42N2O14; molecular weight: 762.76.
- Soluble in DMSO at 125 mg/mL (ultrasonic recommended).
- Available in a 100 MG pack size.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.