Learn More
Medchemexpress LLC HY-116063 10mg , Doxorubicin-SMCC CAS:400647-59-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11606310MG
Encompass_Preferred
HY-116063 10mg Doxorubicin-SMCC CAS: 400647-59-8 Doxorubicin-SMCC is a drug-linker conjugate for ADC. Doxorubicin-SMCC contains a non-cleavable ADC linker and a DNA topoisomerase II inhibitor Doxorubicin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1307