Learn More
Medchemexpress LLC Doxorubicin-SMCC 50mg | 400647-59-8 | 762.76 | C39H42N2O14 | 50 MG

Supplier: Medchemexpress LLC HY11606350MG
Doxorubicin-SMCC is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples the DNA topoisomerase II inhibitor doxorubicin to a non-cleavable SMCC linker. It is supplied as a solid powder and is available in solution formats; intended for research use only.
- Provides non-cleavable SMCC maleimide-thiol conjugation chemistry.
- Contains a doxorubicin payload (DNA topoisomerase II inhibitor).
- High purity, typically about 98.8%.
- Molecular weight ~762.76 g/mol; formula C39H42N2O14.
- Available in multiple pack sizes, common pack 50 mg.
- Store powder at -20°C for long term; store solutions at -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.