Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Doxorubicin-SMCC 5mg | 400647-59-8 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1160635MG

Encompass_Preferred

Doxorubicin-SMCC is an antibody-drug conjugate (ADC) drug-linker conjugate that links the anthracycline topoisomerase II inhibitor doxorubicin to a non-cleavable SMCC linker for research use. It is supplied as a solid or as a DMSO solution and is intended for in vitro ADC research and linker-drug evaluation.

  • Contains non-cleavable SMCC linker
  • Delivers anthracycline topoisomerase II inhibitor doxorubicin
  • High purity suitable for research applications
  • Available as solid and as 10 mM solution in DMSO
  • Soluble in DMSO at up to 125 mg/mL with ultrasonic assistance

Catalog No. 50-002-28084


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.