Learn More
Selleck Chemical LLC Doxycycline S5159-1g
SDP
Supplier:  Selleck Chemical LLC S51591G
Encompass_Preferred
Doxycycline (Vibramycin Doxytetracycline Doxiciclina Doxycyclinum) is an antibiotic that is used in the treatment of a number of types of infections caused by bacteria and protozoa Doxycycline is also a nonspecific matrix metalloproteinase (MMP) inhibitor
Catalog No. 50-311-2451