Learn More
Research Products International Corp Doxycycline Hydrochloride, 5 Grams
SDP
Supplier:  Research Products International Corp D430205.0
Encompass_Preferred
Doxycycline HCl is a widely used antibiotic in the life science marketplace. 
It belongs to the tetracycline class of antibiotics and exhibits broad-spectrum activity against both gram-positive and gram-negative bacteria, as well as certain other microorganisms. 
In the life science marketplace, Doxycycline HCl finds applications in various fields, including research and biotechnology. It is commonly used as an antibacterial agent in cell culture systems, where it helps prevent bacterial contamination in cell lines and maintain sterile conditions. 
Doxycycline HCl is also utilized in molecular biology techniques such as gene expression studies and protein production. It can be added to media to select for cells containing specific genes or recombinant DNA constructs, as it inhibits protein synthesis in bacteria and other organisms.
Catalog No. 50-136-8224