Learn More
Selleck Chemical LLC Doxycycline HCl S4005-50mg
SDP
Supplier:  Selleck Chemical LLC S400550MG
Encompass_Preferred
Doxycycline HCl is a tetracycline antibiotic that commonly used to treat a variety of infections and also a MMP inhibitor
Catalog No. 50-310-9647