Learn More
Med Vet International Doxycycline Hy Capsules (Oracea), 50mg, 50ct
Supplier:  Med Vet International RX72578005418
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Doxycycline is a generic version of Oracea. It belongs to a class of antibiotics known as tetracyclines. It should be stored at room temperature between 59-86 F and away from light and moisture.
Catalog No. 50-282-8147