Learn More
Gold Biotechnology Inc Doxycycline Hyclate 10 g
SDP
Supplier:  Gold Biotechnology Inc D50010
Encompass_Preferred
Doxycycline hyclate is a semisynthetic tetracycline antibiotic effective against bacteria such as Mycoplasma pneumoniae and Haemophilus influenzae It is also a broad-spectrum inhibitor of matrix metalloproteases (MMPs) Doxycycline hyclate is routinely used to select for cells containing the pcDNA3 1 resistance plasmid in cell line 293 at an effective concentration of 2 g/mL It is freely soluble in aqueous solution
Catalog No. 50-005-62085