Learn More
Selleck Chemical LLC Doxycycline Hyclate 100mg 24390-14-5
SDP
Supplier:  Selleck Chemical LLC S4163100MG
Encompass_Preferred
Doxycycline is a member of the tetracycline antibiotics group, and is commonly used to treat a variety of infections. It is also an inhibitor of matrix metallo-proteinases (MMP). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7977