Learn More
Medchemexpress LLC Doxycycline hyclate | 24390-14-5 | 512.94 | 10 G
SDP
Supplier:  Medchemexpress LLC HYN0565B10G
Encompass_Preferred
Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. It shows antibacterial activity and anti-cancer cell proliferation activity. Doxycycline hyclate can be used to construct gene expression regulation models.
  • Orally active and broad-spectrum metalloproteinase (MMP) inhibitor.
  • Possesses antibacterial activity.
  • Shows anti-cancer cell proliferation activity.
  • Applicable for constructing gene expression regulation models.
  • Offers longer activity and faster peak concentration compared to Tetracycline.
Catalog No. 50-003-88775