Learn More
Enzo Life Sciences Doxycycline . hyclate, (5g), CAS Number: 24390-14-5
SDP
Supplier:  Enzo Life Sciences ALX380273G005
Encompass_Preferred
MMP inhibitor. Broad spectrum antibiotic and bacteriostatic. Shows antiprotozoal properties. Potent inhibitor of MMPs (matrix metalloproteinases) in vivo. Inhibits collagen synthesis. Increases and promotes smooth muscle cell adhesion. Inhibits proliferation and migration. Protects microvasculature by inhibiting plasminogen systems. Alternative Name: Doxycycline . hydrochloride . hemiethanolate . hemihydrate. Formula: C46H58Cl2N4O18. MW: 1025.87 g/mol. CAS Number: 24390-14-5. MI: 14: 3440. Purity: ≥98% (HPLC). Appearance: Yellow crystalline solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C
Catalog No. NC0233297