Learn More
Selleck Chemical LLC Doxycycline Hyclate S4163-1g
SDP
Supplier:  Selleck Chemical LLC S41631G
Encompass_Preferred
Doxycycline Hyclate is a member of the tetracycline antibiotics group and is commonly used to treat a variety of infections It is also an inhibitor of matrix metallo-proteinases (MMP)
Catalog No. 50-310-8707