Learn More
Alkali Scientific Doxycycline Hydrochloride, 10 G
SDP
Supplier:  Alkali Scientific DOX002CP
Encompass
Doxycycline Hydrochloride is a tetracycline-class antibiotic used extensively in laboratory and research applications for its broad-spectrum antibacterial activity. It inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, effectively stopping cell growth and replication. The compound is active against a variety of Gram-positive and Gram-negative organisms, making it useful for microbiological assays, contamination control, and antimicrobial susceptibility testing. Due to its high solubility in hydrochloride form, it is easy to prepare and incorporate into culture media or experimental solutions. It is also commonly used in gene expression systems where controlled induction is required.
Catalog No. 50-006-60169