Learn More
Alkali Scientific Doxycycline Hydrochloride, 100 G
Supplier: Alkali Scientific DOX003CP
Doxycycline Hydrochloride is a widely used tetracycline antibiotic known for its broad-spectrum activity against Gram-positive and Gram-negative bacteria. It acts by inhibiting protein synthesis through binding to the bacterial 30S ribosomal subunit, preventing translation and cell growth. In laboratory settings, it is frequently used for antimicrobial research, cell culture contamination control, and preparation of selective media. The hydrochloride salt form provides improved solubility and stability in aqueous solutions. Additionally, doxycycline is commonly used in molecular biology applications such as inducible gene expression systems, where it regulates transcription in controlled experimental models.