Learn More
Alkali Scientific Doxycycline Hydrochloride, 25 G
Supplier: Alkali Scientific DOX004CP
Doxycycline Hydrochloride is a broad-spectrum tetracycline antibiotic used in microbiology and research laboratories for inhibiting bacterial growth. It works by binding to the 30S ribosomal subunit and blocking protein synthesis, preventing bacterial replication. The compound is effective against a wide range of organisms and is commonly used in antimicrobial testing, culture media preparation, and contamination control. Its hydrochloride form enhances solubility, allowing for convenient preparation of working solutions. Doxycycline is also widely used in molecular biology experiments, particularly in inducible gene expression systems where it functions as a regulatory compound.