Learn More
Alkali Scientific Doxycycline Hydrochloride, 250 G
SDP
Supplier:  Alkali Scientific DOX005CP
Encompass_Preferred
Doxycycline Hydrochloride is a tetracycline-class antibiotic widely utilized in microbiology, biotechnology, and molecular research. It inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, effectively preventing cell growth and replication. The compound demonstrates broad-spectrum activity against both Gram-positive and Gram-negative bacteria, making it suitable for antimicrobial assays, culture media preparation, and contamination control in laboratory environments. Its hydrochloride salt form ensures good solubility in aqueous solutions. In addition, doxycycline is frequently used in inducible gene expression systems, where it serves as a regulatory agent to control transcription in experimental studies.
Catalog No. 50-006-60225