Learn More
Alkali Scientific Doxycycline Hydrochloride, 5 G
Supplier: Alkali Scientific DOX006CP
Doxycycline Hydrochloride is a broad-spectrum antibiotic belonging to the tetracycline class, commonly used in laboratory research and microbiological applications. It functions by inhibiting bacterial protein synthesis through binding to the 30S ribosomal subunit, preventing cell growth and replication. The compound is effective against a wide range of bacterial species and is widely used in antimicrobial testing, contamination control, and selective media preparation. Its hydrochloride form enhances solubility, making it easy to prepare aqueous solutions for laboratory use. Doxycycline is also used in molecular biology systems as a regulatory agent in inducible gene expression experiments.