Learn More
Medchemexpress LLC Doxycycline (hydrochloride hemiethanolate hemihydrate) | 24390-14-5 | 512.94 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0565BR
Encompass_Preferred
Doxycycline hyclate is an orally active, broad-spectrum antibiotic and metalloproteinase (MMP) inhibitor. It exhibits antibacterial and anti-cancer cell proliferation activity, and can be used for constructing gene expression regulation models. It is intended for research use only.
  • Affects growth of glioma cells.
  • Decreases MT-CO1 protein content in SVG cells.
  • Reduces proliferation of human cell lines.
  • Inhibits cell viability of breast cancer cells.
  • Reduces MMP-9 activity in mice.
  • Used as an inducer in molecular biology for gene expression control.
Catalog No. 50-003-89194