Learn More
Medchemexpress LLC Doxycycline (monohydrate) | 17086-28-1 | 99.0% | 462.45 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00892310MM/1ML
Encompass_Preferred
Doxycycline monohydrate is an antibiotic and a broad-spectrum metalloproteinase (MMP) inhibitor. It is intended for research use only.
  • Reduces expression of mitochondrial encoded cytochrome c oxidase I (MT-CO1) protein and increases glucose consumption.
  • Protects cells from hypoxia-induced glioma cell death at low concentrations.
  • Reduces MMP-9 activity and collagen degradation.
  • Prevents vascular lesions in heterozygous Col3a1 knockout mouse models.
  • Used as an inducer to control gene expression in Tet-On/Tet-Off systems.
  • Associated with clinical trials for various conditions.
Catalog No. 50-004-51987