Learn More
Apexbio Technology LLC Doxylamine Succinate 562-10-7 50mg
SDP
Supplier:  Apexbio Technology LLC B173550
Encompass_Preferred
Doxylamine succinate (CAS 562-10-7) is a first-generation antihistamine that acts as a competitive antagonist at histamine H1 receptors By blocking these receptors it effectively prevents histaminergic signaling leading to pronounced sedative and anticholinergic effects In biomedical research doxylamine succinate is utilized to investigate histaminergic pathways study the pharmacology of H1 receptor antagonists and explore mechanisms underlying sedation and anticholinergic activity Its well-characterized mode of action makes it a valuable reference compound in studies on allergic responses sleep physiology and receptor-ligand interactions
Catalog No. 50-224-0573