Learn More
Med Vet International Miltex Vantage Doyen Intestinal Hemostatic Forceps, 9in, Curved, Longitudinal Serrations
Supplier:  Med Vet International MILV916162
Encompass_Preferred
The Miltex Vantage V916-162 Doyen intestinal forceps are made from high quality Japanese stainless steel and measure 9" in length. They have a finger ring handle with a ratchet lock and curved, flexible blades with longitudinal serrations
Catalog No. 50-264-3477