Learn More
Apexbio Technology LLC DiscoveryProbe™ DNA Damage/DNA Repair Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Supplier:  Apexbio Technology LLC L10330.096
Encompass_Preferred
DiscoveryProbe DNA Damage/DNA Repair Library comprises 481 chemically diverse compounds selected based on their known involvement in DNA damage recognition signaling and repair pathways Compounds in this collection target biologically relevant DNA repair mechanisms including HDAC modulation ATM/ATR signaling pathways and PARP inhibition Representative compounds encompass established modulators and inhibitors extensively studied in pre-clinical and clinical research approved by regulatory agencies such as FDA This specialized chemical library is suitable for mechanistic and functional studies drug screening assays and cellular assays aimed at understanding DNA repair processes and developing therapeutic interventions in oncology research and related biomedical applications
Catalog No. 50-000-63273
Add to Cart
Request a Quote