Learn More
Apexbio Technology LLC DiscoveryProbe™ MAPK Inhibitor Library 100uL/well(10 mM solution),96 Well Deep Well Plate
SDP
Supplier:  Apexbio Technology LLC L10430.096
Encompass_Preferred
DiscoveryProbe MAPK Inhibitor Library is a curated collection of 117 kinase-targeting compounds with inhibitory activity focused on key members within the mitogen-activated protein kinase (MAPK) signaling cascades including RAF kinases MEK1/2 JNK p38 and VEGFR The MAPK signaling pathway regulates diverse cellular processes such as cell proliferation differentiation stress responses and apoptosis and is frequently targeted in cancer research and drug discovery Representative inhibitors in this collection such as Sorafenib (FDA-approved targeting RAF kinase/VEGFR) and Trametinib (FDA-approved specifically inhibiting MEK1/2) exemplify the library s coverage This integrated toolset allows biomedical researchers to systematically investigate the MAPK signaling network s function and therapeutic targeting potential
Catalog No. 50-000-62824