Learn More
Medchemexpress LLC HY-18973 10mg Medchemexpress, Dp44mT CAS:152095-12-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1897310MG
Encompass_Preferred
Medchemexpress, HY-18973 10mg Dp44mT CAS:152095-12-0 Dp44mT is an iron chelator with selective anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2104