Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco's Phosphate Buffered Saline Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture | MFCD00131855 | 500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D8537500ML
Encompass
Dulbecco's Phosphate Buffered Saline Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture | MFCD00131855 | 500ML
Catalog No. NC2061729