Learn More
Medchemexpress LLC DpC 10mM 1mL | 1382469-39-7 | 1 ML

Supplier: Medchemexpress LLC HY11424310MM 1ML
DpC (di-2-pyridylketone 4-cyclohexyl-4-methyl-3-thiosemicarbazone) is an anti-tumor small molecule provided as a ready-to-use 10 mM solution in DMSO or as a solid for custom preparation. It inhibits cancer cell proliferation (reported IC50 0.007-0.096 μM) and is used in in vitro and in vivo studies.
- Ready-to-use 10 mM solution in DMSO, 1 mL.
- Also available as solid for custom stock preparation.
- High purity (~98.7%) suitable for biological assays.
- High solubility in DMSO (≈62.5 mg/mL) for convenient stock preparation.
- Storage: powder -20°C; prepared solutions -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.