Learn More
Medchemexpress LLC DPC-681 | 284661-68-3 | 98.8% | 669.85 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19400
Encompass_Preferred
DPC-681 is a potent and selective inhibitor of HIV protease. It is designed for research use only and not intended for patient use.
  • Potent and selective inhibitor of HIV protease
  • Active against wild-type HIV-1
  • Retains potency against mutant HIVs with D30N mutation
  • Shows minimal loss of potency against mutant variants with multiple amino acid substitutions
Catalog No. 50-003-84722