Learn More
Hach Company DPD Free Chlorine Reagent Accuvac Ampules, pk/25
Supplier:  Hach Company 2502025
Encompass
For determination of Free Chlorine by the DPD Method, using Accuvac Ampules. USEPA accepted for reporting drinking water analysis using Hach Method 8021. Range: 0.02 - 2.00 mg/L Cl₂. Package of 25 ampules.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6096