Learn More
Hach Company DPD Free Chlorine Reagent Powder Pillows, 10 mL, pk/100
Supplier:  Hach Company 2105569
Encompass
For free chlorine determination by the DPD Free Chlorine method. USEPA accepted for reporting for drinking water analysis using Hach Method 8021. Range: 0.02 - 2.00 mg/L Cl₂. Sample Size: 10 mL. Pack of 100 powder pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5392