Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 25 mL, pk/1000
Supplier:  Hach Company 1406428
Encompass
For total chlorine determination by the DPD method. USEPA accepted for reporting drinking and wastewater analysis using Hach Method 8167. Ranges: Low Range: up to - 2.00 mg/L; Mid Range: 0.05 - 4.00 mg/L; High Range: 0 .1- 10.0 mg/L. Sample Size: Low Range - 25 mL. Mid Range: 10 mL. High Range: 5 mL. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-4909