Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 10 mL, pk/1000
Supplier: Hach Company 2105628
For total chlorine determination by the DPD Method. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Range: 0.02 - 2.00 mg/L. Sample Size: 10 mL. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.