Learn More
Cayman Chemical dPE-PEG 2000-CoH 10mg
SDP
Supplier:  Cayman Chemical 4216410 MG
Encompass_Preferred
A PEGylated derivative of DSPE; has been used to link transferrin receptors to the surface of liposomes encapsulating doxorubicin for targeted delivery of doxorubicin in vitro and in vivo; nanomicelles containing DSPE-PEG(2000)-COOH linked to borneol and encapsulating doxorubicin reduce tumor growth and metastasis in an in situ C6 murine model of glioblastoma
Catalog No. 50-294-5810