Learn More
Cayman Chemical dPE-PolysarcosIn50 25mg
SDP
Supplier:  Cayman Chemical 3904325 MG
Encompass_Preferred
A FASN inhibitor (IC50 0.5 UM) inhibits infection of an infectious-clone-derived SARS-CoV-2 reporter virus in HEK293T cells expressing human ACE2 (EC50 4 nM)
Catalog No. 50-291-9479