Learn More
CPC Scientific H-Tyr-DPen-Gly-(4Cl)Phe-DPen-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ENKP005A
Encompass
Highly selective ligand with high affinity for the delta-opioid receptor.SEQUENCE: H-Tyr-DPen-Gly-(4Cl)Phe-DPen-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Y-DPen-G-(4Cl)Phe-DPenMOLECULAR FORMULA: C30H38N5O7S2Cl1MOLECULAR WEIGHT: 680.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: PainREFERENCES:L.K. Vaughn et al., Life Sciences, 45, 1001 (1989)SKU(s): ENKP-005A, ENKP-005B
Catalog No. 50-210-9256